siRNA Details
| Gene Symbol: LPP-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036497.1 | 1 | 2572 | |||
| GenBank mRNA | BC042414.1 | - | 1 | 2543 | ||
| JX046915.1 | - | 1 | 183 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB539865.1 | - | 1 | 139 | Chr. 3 - 188151207 - 188151655 [-] on Build GRCh38 | |

LPP-AS2
Genomic Map