siRNA Details
| Gene Symbol: LPP-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036497.1 | 1 | 98 | |||
| GenBank mRNA | BC042414.1 | - | 1 | 67 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPP-AS2| Gene Symbol: LPP-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036497.1 | 1 | 98 | |||
| GenBank mRNA | BC042414.1 | - | 1 | 67 | ||