siRNA Details
| Gene Symbol: C1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297721.1 | NP_001284650.1 | 5 | 1339 | ||
| NM_001297723.1 | NP_001284652.1 | 6 | 1441 | |||
| GenBank mRNA | AK290583.1 | - | 6 | 1213 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC017002.1 | - | 863 | Chr. - - [-] on Build | ||

C1orf43
Genomic Map