siRNA Details
| Gene Symbol: SYNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | KU310547.1 | - | 20 | 4889 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC031033.1 | - | 1 | 418 | Chr. 6 - 152433525 - 152434485 [-] on Build GRCh38 | |

SYNE1
Genomic Map