siRNA Details
Gene Symbol: LOC102546226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105021.1 | 5 | 586 | |||
GenBank mRNA | BC043237.1 | - | 5 | 591 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI559941.1 | - | 5 | 591 | Chr. 5 - 88409847 - 88439090 [-] on Build GRCh38 |
