siRNA Details
| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038373.1 | 3 | 191 | |||
| NR_134458.1 | 2 | 371 | ||||
| GenBank mRNA | BC043233.1 | - | 3 | 199 | ||
| HY014641.1 | - | 2 | 374 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI560079.1 | - | 3 | 199 | Chr. 10 - 73247360 - 73274941 [+] on Build GRCh38 | |

DNAJC9-AS1
Genomic Map