siRNA Details
| Gene Symbol: LINC00950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL832569.1 | - | 1 | 1012 | ||
| Gene Symbol: TMEM8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011517916.2 | XP_011516218.1 | 9 | 7313 | ||

LINC00950
Genomic Map