siRNA Details
| Gene Symbol: GATB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK124986.1 | - | 1 | 2466 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK054809.1 | - | 1 | 99 | Chr. 4 - 151670846 - 151673520 [-] on Build GRCh38 | |

GATB
Genomic Map