siRNA Details
| Gene Symbol: LOC101929454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109944.1 | 3 | 864 | |||
| GenBank mRNA | BC002741.2 | - | 3 | 864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929454| Gene Symbol: LOC101929454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109944.1 | 3 | 864 | |||
| GenBank mRNA | BC002741.2 | - | 3 | 864 | ||