siRNA Details
| Gene Symbol: LOC253805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028622.2 | - | 1 | 171 | ||
| BC050316.1 | - | 1 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC253805| Gene Symbol: LOC253805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028622.2 | - | 1 | 171 | ||
| BC050316.1 | - | 1 | 164 | |||