siRNA Details
| Gene Symbol: SFTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY102071.1 | - | 5 | 1088 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000521945.1 | - | 5 | 1088 | Chr. 14 - 36473207 - 36519521 [-] on Build GRCh38 | |

SFTA3
Genomic Map