siRNA Details
| Gene Symbol: LINC01450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110831.1 | 3 | 1158 | |||
| GenBank mRNA | BX647937.1 | - | 3 | 1159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01450| Gene Symbol: LINC01450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110831.1 | 3 | 1158 | |||
| GenBank mRNA | BX647937.1 | - | 3 | 1159 | ||