siRNA Details
| Gene Symbol: LINC01450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110831.1 | 3 | 1468 | |||
| GenBank mRNA | BX647937.1 | - | 3 | 1469 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443406.1 | - | 3 | 1468 | Chr. 7 - 40964667 - 40979939 [-] on Build GRCh38 | |

LINC01450
Genomic Map