siRNA Details
| Gene Symbol: CDC42BPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130237.1 | - | 1 | 847 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM699643.1 | - | 1 | 279 | Chr. 1 - 227060152 - 227060680 [-] on Build GRCh38 | |

CDC42BPA
Genomic Map