siRNA Details
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005250045.2 | XP_005250102.1 | 19 | 3531 | ||
| XM_017012558.1 | XP_016868047.1 | 20 | 3651 | |||
| XM_017012559.1 | XP_016868048.1 | 19 | 3497 | |||
| GenBank mRNA | AK124257.1 | - | 1 | 1102 | ||

BRAF
Genomic Map