siRNA Details
| Gene Symbol: PAK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128166.2 | NP_001121638.1 | 16 | 6392 | ||
| NM_001128167.2 | NP_001121639.1 | 15 | 6319 | |||
| NM_001128168.2 | NP_001121640.1 | 20 | 6641 | |||
| NM_001128172.2 | NP_001121644.1 | 15 | 5994 | |||
| NM_001128173.2 | NP_001121645.1 | 19 | 6578 | |||
| NM_001324325.1 | NP_001311254.1 | 16 | 6470 | |||
| NM_001324326.1 | NP_001311255.1 | 16 | 6315 | |||
| NM_001324327.1 | NP_001311256.1 | 18 | 6427 | |||
| NM_001324328.1 | NP_001311257.1 | 17 | 6360 | |||
| NM_001324329.1 | NP_001311258.1 | 19 | 6575 | |||
| NM_001324330.1 | NP_001311259.1 | 16 | 6467 | |||
| NM_001324331.1 | NP_001311260.1 | 17 | 6382 | |||
| NM_001324332.1 | NP_001311261.1 | 18 | 6533 | |||
| NM_001324333.1 | NP_001311262.1 | 18 | 6508 | |||
| NM_001324334.1 | NP_001311263.1 | 17 | 6463 | |||
| NM_002578.4 | NP_002569.1 | 18 | 6530 | |||
| NR_136747.1 | 18 | 6557 | ||||
| NR_136748.1 | 16 | 6192 | ||||
| XM_017029557.1 | XP_016885046.1 | 17 | 6429 | |||
| XM_017029561.1 | XP_016885050.1 | 18 | 6532 | |||
| XM_017029563.1 | XP_016885052.1 | 20 | 6633 | |||
| GenBank mRNA | AK124702.1 | - | 1 | 2108 | ||

PAK3
Genomic Map