siRNA Details
| Gene Symbol: PAK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128166.2 | NP_001121638.1 | 16 | 6077 | ||
| NM_001128167.2 | NP_001121639.1 | 15 | 6004 | |||
| NM_001128168.2 | NP_001121640.1 | 20 | 6326 | |||
| NM_001128172.2 | NP_001121644.1 | 15 | 5679 | |||
| NM_001128173.2 | NP_001121645.1 | 19 | 6263 | |||
| NM_001324325.1 | NP_001311254.1 | 16 | 6155 | |||
| NM_001324326.1 | NP_001311255.1 | 16 | 6000 | |||
| NM_001324327.1 | NP_001311256.1 | 18 | 6112 | |||
| NM_001324328.1 | NP_001311257.1 | 17 | 6045 | |||
| NM_001324329.1 | NP_001311258.1 | 19 | 6260 | |||
| NM_001324330.1 | NP_001311259.1 | 16 | 6152 | |||
| NM_001324331.1 | NP_001311260.1 | 17 | 6067 | |||
| NM_001324332.1 | NP_001311261.1 | 18 | 6218 | |||
| NM_001324333.1 | NP_001311262.1 | 18 | 6193 | |||
| NM_001324334.1 | NP_001311263.1 | 17 | 6148 | |||
| NM_002578.4 | NP_002569.1 | 18 | 6215 | |||
| NR_136747.1 | 18 | 6242 | ||||
| NR_136748.1 | 16 | 5877 | ||||
| XM_017029557.1 | XP_016885046.1 | 17 | 6114 | |||
| XM_017029561.1 | XP_016885050.1 | 18 | 6217 | |||
| XM_017029563.1 | XP_016885052.1 | 20 | 6318 | |||
| GenBank mRNA | AK124702.1 | - | 1 | 1793 | ||

PAK3
Genomic Map