siRNA Details
| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC045169.2 | - | 1 | 287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC1| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC045169.2 | - | 1 | 287 | ||