siRNA Details
| Gene Symbol: MYO15A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055873.1 | - | 1 | 2490 | ||
| Gene Symbol: DRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI478627.1 | - | 1 | 18 | ||

MYO15A
Genomic Map