siRNA Details
| Gene Symbol: SYNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537552.2 | XP_011535854.1 | 3 | 6258 | ||
| GenBank mRNA | AK095279.1 | - | 1 | 1863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYNPO| Gene Symbol: SYNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537552.2 | XP_011535854.1 | 3 | 6258 | ||
| GenBank mRNA | AK095279.1 | - | 1 | 1863 | ||