siRNA Details
| Gene Symbol: SYNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537552.2 | XP_011535854.1 | 3 | 6747 | ||
| GenBank mRNA | AK095279.1 | - | 1 | 2352 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA164568.1 | - | 1 | 72 | Chr. 5 - 150653868 - 150654224 [+] on Build GRCh38 | |

SYNPO
Genomic Map