siRNA Details
Gene Symbol: LOC101928861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120513.1 | 1 | 279 | |||
GenBank mRNA | BC038554.1 | - | 1 | 285 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120513.1 | 1 | 279 | |||
GenBank mRNA | BC038554.1 | - | 1 | 285 |