siRNA Details
| Gene Symbol: LINC00354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046992.1 | 3 | 193 | |||
| GenBank mRNA | BC032817.1 | - | 1 | 391 | ||
| BX117816.1 | - | 3 | 185 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI520510.1 | - | 392 | Chr. - - [-] on Build | ||

LINC00354
Genomic Map