siRNA Details
| Gene Symbol: LINC01258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110951.1 | 4, 5 | 1136 | |||
| GenBank mRNA | AK057434.1 | - | 4, 5 | 1136 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01258| Gene Symbol: LINC01258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110951.1 | 4, 5 | 1136 | |||
| GenBank mRNA | AK057434.1 | - | 4, 5 | 1136 | ||