siRNA Details
| Gene Symbol: LINC01258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110951.1 | 3 | 426 | |||
| GenBank mRNA | AK057434.1 | - | 3 | 426 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB022894.1 | - | 3 | 426 | Chr. 4 - 38453548 - 38523180 [-] on Build GRCh38 | |

LINC01258
Genomic Map