siRNA Details
| Gene Symbol: KIAA1614-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026359.1 | - | 2 | 1087 | ||
| Gene Symbol: STX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011509058.1 | XP_011507360.1 | 8 | 2006 | ||
| XM_011509059.1 | XP_011507361.1 | 7 | 1781 | |||
| XM_017000006.1 | XP_016855495.1 | 6 | 1615 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415647.1 | - | 2 | 1087 | Chr. 1 - 180949699 - 180954887 [-] on Build GRCh38 | |

KIAA1614-AS1
Genomic Map