siRNA Details
| Gene Symbol: LINC01544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103449.1 | 4 | 1574 | |||
| GenBank mRNA | AK054836.1 | - | 4 | 1574 | ||
| AK091955.1 | - | 1 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01544| Gene Symbol: LINC01544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103449.1 | 4 | 1574 | |||
| GenBank mRNA | AK054836.1 | - | 4 | 1574 | ||
| AK091955.1 | - | 1 | 346 | |||