siRNA Details
Gene Symbol: LINC01544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103449.1 | 4 | 2099 | |||
GenBank mRNA | AK054836.1 | - | 4 | 2099 | ||
AK091955.1 | - | 1 | 871 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103449.1 | 4 | 2099 | |||
GenBank mRNA | AK054836.1 | - | 4 | 2099 | ||
AK091955.1 | - | 1 | 871 |