siRNA Details
Gene Symbol: MKNK1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038403.1 | 8 | 2085 | |||
GenBank mRNA | AK097731.1 | - | 8 | 2085 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MKNK1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038403.1 | 8 | 2085 | |||
GenBank mRNA | AK097731.1 | - | 8 | 2085 |