siRNA Details
| Gene Symbol: NOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF446131.1 | - | 1 | 355 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE297602.1 | - | 1 | 302 | Chr. 12 - 117331433 - 117452170 [-] on Build GRCh38 | |

NOS1
Genomic Map