siRNA Details
| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094870.1 | - | 1 | 994 | ||
| AK128349.1 | - | 1 | 1732 | |||
| AL390144.1 | - | 1 | 409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRF1| Gene Symbol: ZNRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK094870.1 | - | 1 | 994 | ||
| AK128349.1 | - | 1 | 1732 | |||
| AL390144.1 | - | 1 | 409 | |||