siRNA Details
| Gene Symbol: LOC101928992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110043.1 | 1 | 16 | |||
| GenBank mRNA | BC050282.1 | - | 1 | 16 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI460272.1 | - | 1 | 21 | Chr. 3 - 184758510 - 184773159 [-] on Build GRCh38 | |

LOC101928992
Genomic Map