siRNA Details
| Gene Symbol: LOC101928992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110043.1 | 3 | 2500 | |||
| GenBank mRNA | BC050282.1 | - | 3 | 2500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928992| Gene Symbol: LOC101928992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110043.1 | 3 | 2500 | |||
| GenBank mRNA | BC050282.1 | - | 3 | 2500 | ||