siRNA Details
| Gene Symbol: LOC101928135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110817.1 | 4 | 604 | |||
| GenBank mRNA | BC035904.1 | - | 4 | 604 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI918059.1 | - | 4 | 604 | Chr. 3 - 34875992 - 35394023 [-] on Build GRCh38 | |

LOC101928135
Genomic Map