siRNA Details
| Gene Symbol: LZTS1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047509.1 | 3 | 473 | |||
| GenBank mRNA | BC038538.1 | - | 3 | 473 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830862.1 | - | 3 | 471 | Chr. 8 - 20275773 - 20290431 [+] on Build GRCh38 | |

LZTS1-AS1
Genomic Map