siRNA Details
Gene Symbol: RSBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001518.1 | XP_016857007.1 | 3 | 2971 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK001115.1 | - | 1 | 870 | Chr. 1 - 113793699 - 113795643 [-] on Build GRCh38 |
