siRNA Details
| Gene Symbol: ZNF337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027802.1 | XP_016883291.1 | 1 | 155 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK091380.1 | - | 1 | 2689 | Chr. 20 - 25688422 - 25691140 [-] on Build GRCh38 | |

ZNF337
Genomic Map