siRNA Details
Gene Symbol: ENDOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524664.2 | XP_011522966.1 | 1 | 77 | ||
XM_011524667.2 | XP_011522969.1 | 1 | 77 | |||
XM_011524678.2 | XP_011522980.1 | 1 | 77 | |||
XM_017024508.1 | XP_016879997.1 | 1 | 77 | |||
XM_017024509.1 | XP_016879998.1 | 1 | 77 | |||
XM_017024512.1 | XP_016880001.1 | 1 | 77 | |||
XM_017024513.1 | XP_016880002.1 | 1 | 77 | |||
XM_017024520.1 | XP_016880009.1 | 1 | 77 |
