siRNA Details
| Gene Symbol: GRIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005253351.3 | XP_005253408.1 | 4 | 12283 | ||
| XM_011520628.2 | XP_011518930.1 | 14 | 15349 | |||
| XM_011520629.2 | XP_011518931.1 | 15 | 15156 | |||
| XM_017019219.1 | XP_016874708.1 | 14 | 14904 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125440.1 | - | 1 | 213 | Chr. 12 - 13547996 - 13553073 [-] on Build GRCh38 | |

GRIN2B
Genomic Map