siRNA Details
| Gene Symbol: HAND2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136192.1 | 4 | 1682 | |||
| NR_136193.1 | 4 | 2379 | ||||
| NR_136194.1 | 4 | 1849 | ||||
| NR_136197.1 | 4 | 1753 | ||||
| NR_136198.1 | 3 | 1570 | ||||
| NR_136199.1 | 5 | 1892 | ||||
| NR_136200.1 | 4 | 2382 | ||||
| NR_136201.1 | 4 | 2549 | ||||
| GenBank mRNA | AB075501.1 | - | 4 | 2290 | ||
| AK021601.1 | - | 1 | 2029 | |||
| DQ831955.1 | - | 1 | 442 | |||
| DQ831956.1 | - | 1 | 442 | |||
| EF529495.1 | - | 4 | 682 | |||
| EF529496.1 | - | 4 | 605 | |||
| EF529497.1 | - | 4 | 553 | |||
| EF529498.1 | - | 4 | 817 | |||
| EF529506.1 | - | 4 | 682 | |||
| EF529507.1 | - | 4 | 605 | |||
| EF529508.1 | - | 4 | 682 | |||
| EF529509.1 | - | 4 | 605 | |||
| EU000853.1 | - | 4 | 1595 | |||
| EU000854.1 | - | 4 | 1595 | |||
| EU000855.1 | - | 3 | 2687 | |||
| EU000856.1 | - | 3 | 2687 | |||

HAND2-AS1
Genomic Map