siRNA Details
| Gene Symbol: KCNK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104210.1 | 4 | 2965 | |||
| XM_011517101.2 | XP_011515403.1 | 4 | 2720 | |||
| XM_011517102.2 | XP_011515404.1 | 4 | 4916 | |||
| XM_011517103.2 | XP_011515405.1 | 4 | 2464 | |||
| XM_017013530.1 | XP_016869019.1 | 4 | 4632 | |||
| XM_017013531.1 | XP_016869020.1 | 4 | 2731 | |||
| GenBank mRNA | AK090707.1 | - | 3 | 2398 | ||
| AK094401.1 | - | 1 | 1961 | |||
| BC034734.1 | - | 3 | 1811 | |||

KCNK9
Genomic Map