siRNA Details
| Gene Symbol: LOC101927580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110917.1 | 4 | 5939 | |||
| GenBank mRNA | CR936694.1 | - | 4 | 5937 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927580| Gene Symbol: LOC101927580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110917.1 | 4 | 5939 | |||
| GenBank mRNA | CR936694.1 | - | 4 | 5937 | ||