siRNA Details
| Gene Symbol: PCDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016580.3 | NP_057664.1 | 4 | 6261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCDH12| Gene Symbol: PCDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016580.3 | NP_057664.1 | 4 | 6261 | ||