siRNA Details
| Gene Symbol: LOC146513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038438.1 | 3 | 1995 | |||
| GenBank mRNA | BC043214.2 | - | 3 | 1999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC146513| Gene Symbol: LOC146513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038438.1 | 3 | 1995 | |||
| GenBank mRNA | BC043214.2 | - | 3 | 1999 | ||