siRNA Details
Gene Symbol: DEFA11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073421.2 | 2 | 212 | |||
GenBank mRNA | AY746436.1 | - | 2 | 212 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DEFA11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073421.2 | 2 | 212 | |||
GenBank mRNA | AY746436.1 | - | 2 | 212 |