siRNA Details
| Gene Symbol: RIMBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX641152.1 | - | 1 | 5342 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX648713.1 | - | 1 | 1924 | Chr. 4 - 161833253 - 161836978 [-] on Build GRCh38 | |

RIMBP2
Genomic Map