siRNA Details
| Gene Symbol: GLIPR1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270396.1 | NP_001257325.1 | 6 | 1640 | ||
| NR_072995.1 | 7 | 1770 | ||||
| XM_011537949.2 | XP_011536251.1 | 6 | 1554 | |||
| XM_011537950.2 | XP_011536252.1 | 6 | 1526 | |||
| XM_017018848.1 | XP_016874337.1 | 7 | 1630 | |||
| XM_017018849.1 | XP_016874338.1 | 7 | 1656 | |||
| GenBank mRNA | AK093175.1 | - | 6 | 1631 | ||
| BC032022.1 | - | 1 | 251 | |||
| BX640916.1 | - | 7 | 1739 | |||

GLIPR1L2
Genomic Map