siRNA Details
| Gene Symbol: LOC105372526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023793.1 | - | 1 | 803 | ||
| Gene Symbol: JAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027707.1 | XP_016883196.1 | 1 | 803 | ||

LOC105372526
Genomic Map