siRNA Details
| Gene Symbol: LOC100419583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125376.1 | 2 | 1907 | |||
| GenBank mRNA | BC031266.1 | - | 2 | 1773 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100419583| Gene Symbol: LOC100419583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125376.1 | 2 | 1907 | |||
| GenBank mRNA | BC031266.1 | - | 2 | 1773 | ||