siRNA Details
| Gene Symbol: STK24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020795.1 | XP_016876284.1 | 1 | 103 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130723.1 | - | 1 | 2649 | Chr. 13 - 98551463 - 98554368 [-] on Build GRCh38 | |

STK24
Genomic Map